missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL2/MCP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
200.00€ - 450.00€
Specifications
| Antigen | CCL2/MCP1 |
|---|---|
| Concentration | 1.42 mg/mL |
| Dilution | Western Blot 0.5 ug/ml, Immunohistochemistry (Negative), ICC/IF (Negative) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18228520
|
Novus Biologicals
NBP1-07034SS |
0.025 mL |
200.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18787843
|
Novus Biologicals
NBP1-07034 |
0.1 mL |
450.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL2/MCP1 Polyclonal specifically detects CCL2/MCP1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry (Negative), ICC/IF (Negative).Specifications
| CCL2/MCP1 | |
| Western Blot 0.5 ug/ml, Immunohistochemistry (Negative), ICC/IF (Negative) | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| 6347 | |
| Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot# P14844] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 16 kDa |
| 1.42 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF | |
| CCL2 | |
| IgG | |
| Affinity Purified | |
| Human and Rat. Mouse reactivity reported by a customer review. Immunogen sequence has 95% homology to mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title