Learn More
Invitrogen™ CCM2L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552561
Description
Immunogen sequence: IRRLVFPKAG RRAACRSSVS RRPLHSMPLY PPDYLIDPQI LLCDYLEKEV KFLGHLTWVT SSLNPSSRDE LLQLLDTARQ LKELPLKTTA EQDSILSLSA RCLLLTWRDN EELILRIPTH E Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
CCM2L gene ontology annotations related to this gene include cardiac atrium morphogenesis; cardiac muscle tissue growth; negative regulation of homotypic cell-cell adhesion; negative regulation of protein binding; positive regulation of fibroblast growth factor production; ventricular trabecula myocardium morphogenesis; wound healing.
Specifications
| CCM2L | |
| Polyclonal | |
| Unconjugated | |
| CCM2l | |
| C20orf160; C24H20orf160; CCM2 like scaffold protein; CCM2 like scaffolding protein; CCM2l; CCM2-like; ccml2; cerebral cavernous malformation 2; cerebral cavernous malformation 2-like; cerebral cavernous malformations 2 protein-like; dJ310O13.5; LOW QUALITY PROTEIN: cerebral cavernous malformations 2 protein-like; RGD1305202; si:dkey-223d9.2; uncharacterized protein C20orf160 homolog; cerebral cavernous malformations 2 protein-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140706 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUG4 | |
| CCM2l | |
| Recombinant protein corresponding to Human CCM2L. Recombinant protein control fragment (Product #RP-89709). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.