Learn More
CCNC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26464-1-AP-20UL
 View more versions of this product
Product Code. 16804925
  • 145.00€ / 20µL
Description
The protein encoded by this gene is a member of the cyclin family of proteins. The encoded protein interacts with cyclin-dependent kinase 8 and induces the phophorylation of the carboxy-terminal domain of the large subunit of RNA polymerase II. The level of mRNAs for this gene peaks in the G1 phase of the cell cycle. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
CCNC
Polyclonal
Unconjugated
CCNC
CCNC, CycC, cyclin C, hSRB11, SRB11 homolog
Rabbit
Antigen Affinity Chromatography
RUO
51813, 892
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P24863, Q62447
Ccnc
CCNC Fusion Protein Ag23955
20 μL
Primary
Human, Mouse