missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
226.00€ - 522.00€
Specifications
| Antigen | CCR1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 5-10 ul/test, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 15860797), Flow (Cell Surface) reported in scientific literature (PMID 20371596), Flow (Intracellular) reported in scientific literature (PMID 20371596) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18798403
|
Novus Biologicals
NB100-56334SS |
0.025 mg |
226.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18000074
|
Novus Biologicals
NB100-56334 |
0.1 mg |
522.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR1 Polyclonal specifically detects CCR1 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular).Specifications
| CCR1 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 5-10 ul/test, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 15860797), Flow (Cell Surface) reported in scientific literature (PMID 20371596), Flow (Intracellular) reported in scientific literature (PMID 20371596) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| NP_001286.1 | |
| 1230 | |
| This antibody was raised against a synthetic peptide of human CCR1 protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| CCR1 | |
| IgG | |
| Immunogen affinity purified | |
| In A375, a 42 kDa band should be observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title