Learn More
CCR1 Antibody (53504R) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB145RJF635
Product Code. 30165820
  • 595.00€ / 0.10mL
Description
CCR1 Monoclonal antibody specifically detects CCR1 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCR1
Monoclonal
Janelia Fluor 635
C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R
Swiss 3T3 mouse cell line transfected with human CCR1, Accession # P32246
RUO
Primary
Human
Liquid
Flow Cytometry, CyTOF
53504R
50mM Sodium Borate
Mouse
0.1 mL
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
1230
Store at 4°C in the dark.
IgG2b