Learn More
CCR6 Mouse anti-Human, Unconjugated, Clone: 1C11A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66801-1-IG-20UL
Description
The CCR6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCR6. This antibody recognizes human antigen. The CCR6 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| CCR6 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunohistochemistry, Immunofluorescence | |
| 1C11A9 | |
| Unconjugated | |
| 20 μL | |
| 1235 | |
| IgG2a |