missing translation for 'onlineSavingsMsg'
Learn More

CCR7 (CD197) Mouse anti-Human , BV786, Clone: 2-L1-A, BD Biosciences™

Mouse Monoclonal Antibody

222.75€ - 492.31€

Specifications

Antigen CCR7 (CD197)
Clone 2-L1-A
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Violet 786
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17267584
View Documents
BD Biosciences
566759
25 Tests
ONLINE EXCLUSIVE PRICE
224.00€
222.75€ / 25 tests
Save 1.25€
0.56% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17257584
View Documents
BD Biosciences
566758
100 Tests
ONLINE EXCLUSIVE PRICE
493.00€
492.31€ / 100 tests
Save 0.69€
0.14% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CCR7 (CD197)
Flow Cytometry
Brilliant Violet 786
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CCR7 Transfected Cell Line
Primary
2-L1-A
Monoclonal
Mouse
Human
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV786 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV786 were removed.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.