Learn More
CCT3 Antibody (8C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42938
This item is not returnable.
View return policy
Description
CCT3 Monoclonal antibody specifically detects CCT3 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| CCT3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 8C8 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/ Immunofluorescence 1:20 - 1:200 | |
| Cctg, chaperonin containing TCP1, subunit 3 (gamma), hTRiC5, TCP1 (t-complex-1) ring complex, polypeptide 5, TRIC5gamma subunit | |
| chaperonin containing TCP1, subunit 3(gamma) - (Uniprot# P49368) | |
| 100 μg | |
| Stem Cell Markers | |
| 7203 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |