missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 (Integrin alpha E) Monoclonal Antibody (B-Ly7), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 62-1038-41
This item is not returnable.
View return policy
Description
Mouse Monoclonal Antibody
CD103 (Integrin alpha E) Monoclonal antibody specifically detects CD103 (Integrin alpha E) in Human samples. It is validated for Flow Cytometry
Specifications
| CD103 (Integrin alpha E) | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P38570 | |
| Itgae | |
| Affinity chromatography | |
| RUO | |
| 3682 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| B-Ly7 | |
| Super Bright 436 | |
| Itgae | |
| A530055J10; alpha-E1; alpha-M290; aM290; CD103; DC; DCs; HML-1 antigen; HUMINAE; Integrin alpha M290; Integrin alpha-E; Integrin alpha-IEL; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction