Learn More
CD104 Rat anti-Human, Brilliant Violet 786, Clone: 439-9B, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 744155
Description
The 439-9B monoclonal antibody specifically recognizes CD104, integrinβ4 chain, a 205 kDa transmembrane glycoprotein, which associates with CD49f (integrinα6 chain) to form theα6/β4 (CD49f/CD104) complex. It is expressed on epithelial cells, Schwann cells, and some tumor cells. The CD49f/CD104 complex is located in the hemidesmosomes of the epidermis, suggesting its role in epidermal cell-basement membrane adhesion. The clone 439-9B was clustered as CD104 at the fifth Human Leucocyte Differentiation Antigen International Workshop. It may be used for immunoprecipitation, immunoblotting and immunohistochemistry on frozen tissue sections.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
Specifications
| CD104 | |
| 439-9B | |
| Brilliant Violet 786 | |
| Integrin β4 chain, Integrin beta 4, ITGB4, GP150, TSP-180 | |
| Human CD104 Protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.