Learn More
CD11a Mouse anti-Human, Brilliant Violet 421, Clone: G43-25B, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 743330
Description
The G43-25B monoclonal antibody specifically binds to CD11a. CD11a is the 180 kD integrin chain which associates with CD18 (integrin 2) to form the heterodimer glycoprotein CD11a/CD18. This is known as the lymphocyte function associated antigen-1 (LFA-1), which is on all leukocytes; and is an adhesion molecule involved in lymphocyte and granulocyte functions. LFA-1 mediates adhesion of lymphoid cells to the vascular endothelium in association with its ligand, the intracellular adhesion molecule-1 (ICAM-1), CD54. Other ligands are ICAM-2 (CD102) and ICAM-3 (CD50).
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
| CD11a | |
| G43-25B | |
| Brilliant Violet 421 | |
| LFA-1α, LFA-1A, LFA1A, Integrin alpha-L, ITGAL | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.