missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b Mouse, anti-Human, FITC, Clone: ICRF44, STEMCELL Technologies
Mouse Monoclonal Antibody
193.05€ - 410.40€
Specifications
| Antigen | CD11b |
|---|---|
| Clone | ICRF44 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17188721
|
Stemcell Technologies
60040FI.1 |
25 Tests |
193.05€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17178721
|
Stemcell Technologies
60040FI |
100 Tests |
410.40€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The ICRF44 antibody reacts with an extracellular epitope on CD11b (integrin αM), an ∼170 kDa type 1 transmembrane glycoprotein which associates non-covalently with CD18 to form the heterodimeric Mac-1 receptor
- Through its interactions with ligands such as ICAM-1 (CD54), ICAM-2 (CD102), ICAM-4 (CD242), iC3b, heparin, and fibrinogen, Mac-1 influences several processes, including the adherence of neutrophils and monocytes to stimulated endothelium, and phagocytosis of complement-coated particles
- CD11b is expressed on the surface of granulocytes, monocytes, NK cells, dendritic cells, tissue macrophages, and subsets of T and B cells, and has been used as a marker to distinguish naive and memory CD8+ T cells
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human Monocyte Enrichment Kit (Catalog #19059), and for labeling human mesenchymal cells grown in MesenCult™ Proliferation Kit (Human; Catalog #05411)
Specifications
| CD11b | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FITC | |
| Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| Primary |
| ICRF44 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title