missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse, FITC, Clone: 3.9, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB24-F-100Tests

 View more versions of this product

Product Code. 15921718

  • 196.35€ / 100 tests

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CD11c
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
ITGAX
Protein A/G Purified
RUO
3687
IgG1 κ
Flow Cytometry
3.9
FITC
XP_011544154
Mouse
100 Tests
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.