missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD134/OX40 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20006-1-AP-20UL
This item is not returnable.
View return policy
Description
Tumor necrosis factor receptor superfamily, member4, also known as ACT35 or CD134 is a cell surface glycoprotein that was discovered through the production of a monoclonal antibody raised against the HUT-102 cell line. It belongs to the tumor necrosis factor receptor superfamily. CD134 was mapped to 1p36 by fluorescence in situ hybridization. CD134 is the primary receptor for feline immunodeficiency virus. CD134 expression can promote viral binding and renders cells permissive for viral entry, productive infection, and syncytium formation. Stimulating the receptor can improve the response to a powerful virus vector and may be useful in vaccine development.Specifications
| CD134/OX40 | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF4 | |
| ACT35, ACT35 antigen, CD134, OX40, OX40L receptor, TNFRSF4, TXGP1L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7293 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43489 | |
| TNFRSF4 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction