Learn More
CD14 Mouse anti-Human, PE, Clone: M5E2, BD
Mouse Monoclonal Antibody
77.19€ - 334.27€
Specifications
| Antigen | CD14 |
|---|---|
| Clone | M5E2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15821259
|
BD Biosciences
561707 |
25 Tests |
77.20€ 77.19€ / Each Save 0.01€ 0.01% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15818138
|
BD Biosciences
555398 |
100 Tests |
335.00€ 334.27€ / Each Save 0.73€ 0.22% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.
Specifications
| CD14 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified |
| M5E2 | |
| Monoclonal | |
| Mouse | |
| Baboon, Canine, Cynomolgus, Human, Rhesus Monkey | |
| Human CD14 Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.