missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD157 Monoclonal Antibody (eBioSY11B5 (SY11B5)), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
178.00€ - 392.00€
Specifications
| Antigen | CD157 |
|---|---|
| Clone | eBioSY11B5 (SY11B5) |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056170
|
Invitrogen™
H041T03B08-A |
100 Tests |
392.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056078
|
Invitrogen™
H041T02B08-A |
25 Tests |
178.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD157 Monoclonal antibody specifically detects CD157 in Human samples. It is validated for Flow CytometrySpecifications
| CD157 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q10588 | |
| 683 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBioSY11B5 (SY11B5) | |
| Flow Cytometry | |
| NovaFluor Blue 660-120S | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase | |
| IgG1 κ | |
| Affinity chromatography | |
| Bst1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title