missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD16 Monoclonal Antibody (3G8), NovaFluor™ Blue 530, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
196.00€ - 418.00€
Specifications
| Antigen | CD16 |
|---|---|
| Clone | 3G8 |
| Concentration | 4 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056149
|
Invitrogen
H006T03B02-A |
100 Tests |
418.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056057
|
Invitrogen
H006T02B02-A |
25 Tests |
196.00€
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD16 Monoclonal antibody specifically detects CD16 in Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow CytometrySpecifications
| CD16 | |
| 4 μL/Test | |
| Monoclonal | |
| Mouse | |
| Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey | |
| A3RFZ7, O75015, P08637 | |
| 2214, 2215, 720006 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
| 3G8 | |
| Flow Cytometry | |
| NovaFluor Blue 530 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 4833442P21Rik, CD16, CD16-2, CD16A, CD16a antigen, CD16b, CD32, CD32 receptor 2, CDw32, cytolytic trigger molecule G7, Fc fragment of IgG intermediate affinity IV receptor, Fc fragment of IgG low affinity IIIa receptor, Fc fragment of IgG receptor IIIa, Fc fragment of IgG receptor IIIb, Fc fragment of IgG, low affinity III, receptor for (CD16), Fc fragment of IgG, low affinity IIIa, receptor, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc fragment of IgG, low affinity III, receptor for (CD16), Fc fragment of IgG, low affinity IIIb, receptor (CD16b), Fc gamma receptor III, Fc gamma receptor IIIa, Fc gamma receptor III-A, Fc gamma receptor IIIb, Fc gamma RIIIa, Fc gammaRIV, Fc receptor, IgG, low affinity III, Fc receptor, IgG, low affinity IV, Fc receptor-like 3, Fcg receptor III, FCG2, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RII-b, Fc-gamma RII-c, fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, fc-gamma RIIIb, fc-gamma RIII-beta, Fc-gamma-RIIb, Fc-gamma-RIIc, FcgammaRIII, FcgammaRIII a.1, FcgammaRIII a.2, FcgammaRIII a.3, FcgammaRIIIA, FcgammaRIV, FCGR2, FCGR2A, FCGR2B, FCGR2C, FCGR3, Fcgr3a, FCGR3B, Fcgr4, FCGRIII, FcgRIV, FcR-10, FcRII-b, FcRII-c, FcRIII, FCRIIIA, FCRIIIb, Fcrl3, IGFR3, IgG Fc gamma receptor III, igG Fc receptor III, IgG Fc receptor III-1, igG Fc receptor III-2, IMD20, immunoglobulin G Fc receptor III, LOC100911825, Low affinity immunoglobulin gamma Fc region receptor III, low affinity immunoglobulin gamma Fc region receptor III-A, Low affinity immunoglobulin gamma Fc region receptor III-B, low affinity immunoglobulin gamma Fc region receptor III-like, Low affinity immunoglobulin gamma Fc region receptor IV, neutrophil-specific antigen NA, RP11-5K23.1, transmembrane receptor FcgammaRIII-X | |
| IgG1 | |
| Affinity chromatography | |
| FCGR3B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title