missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD160 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00€ - 551.00€
Specifications
| Antigen | CD160 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653259
|
Novus Biologicals
NBP2-41234-0.025mg |
0.025 mg |
195.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18148568
|
Novus Biologicals
NBP2-41234 |
0.1 mg |
551.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD160 Polyclonal specifically detects CD160 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| CD160 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| BY55FLJ46513, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55 | |
| CD160 | |
| IgG | |
| Affinity Purified | |
| 25 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Immunology, Innate Immunity, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 11126 | |
| Antibody was raised against a 17 amino acid peptide near the center of human CD160. The immunogen is located within amino acids 50 - 100 of CD160 . Amino Acid Squence: SPETSLKQLRLKRDPG | |
| Primary | |
| Multiple isoforms of CD160 are known to exist. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title