Learn More
CD161 Monoclonal Antibody (10/78), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
301.00€
Specifications
Antigen | CD161 |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-eFluor 710 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15381290
|
Affymetrix eBioscience
46-1610-82 |
100 μg |
301.00€
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Plays an inhibitory role on natural killer (NK) cells cytotoxicity. Activation results in specific acid sphingomyelinase/SMPD1 stimulation with subsequent marked elevation of intracellular ceramide. Activation also leads to AKT1/PKB and RPS6KA1/RSK1 kinases stimulation as well as markedly enhanced T-cell proliferation induced by anti-CD3. Acts as a lectin that binds to the terminal carbohydrate Gal-alpha(1,3)Gal epitope as well as to the N-acetyllactosamine epitope. Binds also to CLEC2D/LLT1 as a ligand and inhibits NK cell-mediated cytotoxicity as well as interferon-gamma secretion in target cells. {ECO:0000269; PubMed:16455998, ECO:0000269; PubMed:16925668, ECO:0000269; PubMed:8077657}.
CD161 Monoclonal antibody specifically detects CD161 in Rat samples. It is validated for Flow Cytometry.Specifications
CD161 | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 κ | |
Affinity chromatography |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Rat | |
Primary | |
Antibody |
For Research Use Only.