missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Mouse, PE-Cy7, Clone: PK136, enQuireBio™
Mouse Monoclonal Antibody
183.75€ - 394.80€
Specifications
| Antigen | CD161 |
|---|---|
| Clone | PK136 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15932488
|
EnquireBio
QAB78-PE7-25ug |
25 μg |
183.75€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15922488
|
EnquireBio
QAB78-PE7-100ug |
100 μg |
394.80€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD161 | |
| 0.5mg/mL | |
| Monoclonal | |
| Mouse | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| 17059 | |
| IgG2a κ | |
| Protein A/G Purified |
| PK136 | |
| Flow Cytometry | |
| PE-Cyanine7 | |
| RUO | |
| NP_001153376 | |
| Klrb1c | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title