missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD175 Monoclonal Antibody (BRIC111)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA180055
This item is not returnable.
View return policy
Description
MA1-80055 recognizes Tn antigen on glycophorin A and glycophorin B in human erythrocytes. A suggested positive control for immunohistochemical applications is breast carcinoma.
Tn, recently designated CD175, is a cryptantigen associated with polyagglutination. It is not expressed on normal haemopoietic cells.
Specifications
| CD175 | |
| Monoclonal | |
| 1 mg/mL | |
| Tris with glycine and <0.1% sodium azide; pH 7.4 | |
| Mouse | |
| Purified | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Hemagglutination Assay, Immunohistochemistry (Frozen) | |
| BRIC111 | |
| Unconjugated | |
| CD175s | |
| Human Tn red blood cells. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction