missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD183 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: 1C6/CXCR3 (also known as 1C6, LS177-1C6), BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566532
This item is not returnable.
View return policy
Description
CD183 Monoclonal antibody specifically detects CD183 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.
Specifications
| CD183 | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 100 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
| Flow Cytometry | |
| 1C6/CXCR3 (also known as 1C6, LS177-1C6) | |
| Mouse | |
| RUO | |
| Human, Baboon, Cynomolgus, Rhesus Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction