missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD19 Monoclonal Antibody (eBio1D3 (1D3)), NovaFluor™ Blue 760, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00€ - 344.00€
Specifications
| Antigen | CD19 |
|---|---|
| Clone | eBio1D3 (1D3) |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30056322
|
Invitrogen™
M004T03B11-A |
100 μg |
344.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30056340
|
Invitrogen™
M004T02B11-A |
25 μg |
178.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD19 Monoclonal antibody specifically detects CD19 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD19 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P25918 | |
| 12478 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| eBio1D3 (1D3) | |
| Flow Cytometry | |
| NovaFluor Blue 760 | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
| IgG2a κ | |
| Affinity chromatography | |
| Cd19 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title