Learn More
CD19 Rat, anti-Mouse, CoraLite Plus 405, Clone: 1D3, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL405-65290-100UG
Description
The CD19 antibody from Proteintech is a rat monoclonal antibody generated with mouse cd19 transfected cell line of mouse CD19. This antibody recognizes mouse antigen. The CD19 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 405.
Specifications
| CD19 | |
| 1D3 | |
| CoraLite Plus 405 | |
| Rat | |
| Primary | |
| Mouse | |
| 150 kDa | |
| IgG2a κ |
| Flow Cytometry | |
| 0.5 mg/ml | |
| AW495831, Cd19, CD19 antigen | |
| 100 μg | |
| 12478 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.