missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a Antibody (SPM120) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45342
This item is not returnable.
View return policy
Description
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CD1a | |
| Monoclonal | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| SPM120 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 0.5 - 1.0 μg/mL | |
| CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
| Human thymus cells | |
| 7 mL | |
| Dendritic Cell Markers, Immunology | |
| 909 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction