missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1c/BDCA-1 Mouse anti-Human, Clone: CD1C/1603, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07517-100ug
This item is not returnable.
View return policy
Description
CD1c/BDCA-1 Monoclonal specifically detects CD1c/BDCA-1 in Human samples. It is validated for ELISA.
Specifications
| CD1c/BDCA-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA | |
| BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
| Recombinant full-length human CD1c/BDCA-1 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| CD1C/1603 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 911 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction