missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2 Antibody (LFA2/8681R), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24030IR
This item is not returnable.
View return policy
Description
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD2 | |
| Monoclonal | |
| DyLight 755 | |
| CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
| Recombinant fragment corresponding to N-terminal of human CD2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells | |
| 914 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| LFA2/8681R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction