missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD2 Monoclonal Antibody (RM2-5), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
179.00€ - 345.00€
Specifications
| Antigen | CD2 |
|---|---|
| Clone | RM2-5 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30087833
|
Invitrogen™
M058T02R03-A |
25 μg |
179.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088026
|
Invitrogen™
M058T03R03-A |
100 μg |
345.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD2 Monoclonal antibody specifically detects CD2 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD2 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Cd2; CD2 antigen; CD2 antigen (p50), sheep red blood cell receptor; CD2 molecule; CD2R; cluster of differentiation 2; Erythrocyte receptor; FLJ46032; LFA2; LFA-2; LFA-3 receptor; Ly37; Ly-37; lymphocyte antigen 37; OX34; OX-34; OX-34 antigen; OX-45 surface antigen homolog to human T lymphocyte CD2 antigen; Rosette receptor; SRBC; surface immunoglobulin superfamily molecule; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5 | |
| IgG2b λ | |
| Affinity chromatography | |
| Antibody |
| RM2-5 | |
| Flow Cytometry | |
| NovaFluor Red 700 | |
| Rat | |
| Mouse | |
| P08920 | |
| 12481 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title