Learn More
CD202b (TIE2) Monoclonal Antibody (TEK4), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
2985.00€ - 14910.00€
Specifications
| Antigen | CD202b (TIE2) |
|---|---|
| Clone | TEK4 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
Description
The TEK4 MAb reacts with mouse Tie-2, also known as CD202. A member of the tyrosine kinase receptor family, Tie-2 is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers^-, c-Kit^+, Sca1^+ cells. Long-term multilineage repopulating cells were detected in Tie-2^+, Lineage^-, c-Kit^+, Sca1^+ cells but not in Tie-2^-, Lineage^-, c-Kit^+, Sca1^+ cells.
CD202b (TIE2) Monoclonal antibody specifically detects CD202b (TIE2) in Mouse samples. It is validated for Control, Flow Cytometry, Functional AssaySpecifications
| CD202b (TIE2) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| AA517024; Angiopoietin-1 receptor; CD202b; CD202b antigen; EC 2.7.10; EC 2.7.10.1; Endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; endothelium-specific receptor tyrosine kinase 2; endothelium-specific receptor tyrosine kinase Tie-2; hTIE2; Hyk; LOC100356675; mTIE2; p140 TEK; protein-tyrosine kinase Tie2; STK1; Tek; TEK receptor tyrosine kinase; TEK tyrosine kinase, endothelial; TEKT1; Tektin-1; tie2; Tie-2; Tunica interna endothelial cell kinase; tyrosine kinase with Ig and EGF homology domains-2; tyrosine kinases that contain immunoglobulin-like loops and epidermal growth factor-similar domains 2; tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor Tie-2; VMCM; VMCM1 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| TEK4 | |
| Flow Cytometry, Functional Assay | |
| Functional Grade | |
| Rat | |
| Mouse | |
| Q02858 | |
| 21687 | |
| Primary | |
| 4° C | |
| TEK |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.