Learn More
CD205 Monoclonal Antibody (205yekta), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen M051T03R03
Description
The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells. Applications Tested: This 205yekta antibody has been tested by flow cytometric analyis of mouse bone marrow cells. This can be used at less than or equal to 0.2 ug per test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100 uL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. NovaFluor dyes are not compatible with DNA intercalating viability dyes. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. Use 5uL of CellBlox Blocking Buffer per stained cell sample containing 10^3 to 10^8 cells. Excitation: 639 nm; Emission: 700 nm; Laser: 633-640 nm (Red) Laser
CD205 (LY25, Lymphocyte Antigen 75) is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 acts as an endocytic receptor that directs captured antigens to the extracellular space towards a specialized antigen processing region. Further, CD205 also prompts the reduced proliferation of B lymphocytes. Diseases associated with CD205 dysfunction include pneumonic plague and lipid pneumonia.
Specifications
| CD205 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q60767 | |
| LY75 | |
| Affinity chromatography | |
| RUO | |
| 17076 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IgG2a κ |
| Flow Cytometry | |
| 205yekta | |
| NovaFluor Red 700 | |
| LY75 | |
| CA-IX; CD205; CLEC13B; C-type lectin domain family 13 member B; DCL-1 fusion; DEC205; DEC-205; DEC-205/CD205 protein; gp200; gp200-MR6; group VI C-type animal lectin; LY75; ly-75; Lymphocyte antigen 75; membrane antigen MN; P54/58N; pMW1; RGD1565962; RP23-10L17.4 | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.