missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD207 (Langerin) Monoclonal Antibody (1C5A1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548742
This item is not returnable.
View return policy
Description
CD207 (Langerin) Monoclonal Antibody for Western Blot, IHC, Flow, ELISA
Langerin (CD207) is a type II membrane-associated C-type lectin known to be expressed exclusively by Langerhans cells. Lamgerin recognizes mannose residues via its single carbohydrate recognition domain (CRD). Langerin is localized not only on the cell surface, but also intracellularly in close association with Birbeck granules. Transfection of Langerin cDNA into fibroblasts creates a compact network of membrane structures with typical features of Birbeck granules (BG). Langerin is thus a potent inducer of membrane superimposition and zippering leading to BG formation.
Specifications
| CD207 (Langerin) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9UJ71 | |
| CD207 | |
| Purified recombinant fragment of human CD207 (AA: extra 155-328) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
| 1C5A1 | |
| Unconjugated | |
| CD207 | |
| CD 207 antigen; CD207; CD207 antigen; CD207 antigen, langerin; CD207 molecule; CD207 molecule, langerin; CLEC4K; C-type lectin domain family 4 member K; C-type lectin domain family 4, member K; Langerhans cell specific c-type lectin; langerin; RGD1565913 | |
| Mouse | |
| Protein G | |
| RUO | |
| 50489 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction