missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD235ab (Glycophorin A/B) Mouse, anti-Human, APC, Clone: HIR2 (GA-R2), STEMCELL Technologies
Mouse Monoclonal Antibody
POA - 589.95€
Specifications
| Antigen | CD235ab (Glycophorin A/B) |
|---|---|
| Clone | HIR2 (GA-R2) |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17149101
|
Stemcell Technologies
60111AZ.1 |
25 μg | |||||||
17139101
|
Stemcell Technologies
60111AZ |
100 μg |
589.95€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The HIR2 (GA-R2) antibody recognizes an epitope common to the N-terminal region of human CD235a (glycophorin A) and CD235b (glycophorin B), homologous type I sialoglycoproteins of the erythrocyte (red blood cell) membrane that bear the antigenic determinants for the MN and Ss blood groups
- CD235ab is expressed on early- and late-stage erythroblasts, mature erythrocytes, and erythroid cell lines such as K562 and HEL, but not on other cell types
- CD235a is abundantly expressed whereas CD235b is a relatively minor membrane component
- The proteins are believed to provide a large mucin-like surface to erythrocytes that acts to minimize aggregation in the circulation
- The HIR2 antibody binds with higher affinity to CD235a than CD235b, and agglutinates untreated erythrocytes
Specifications
| CD235ab (Glycophorin A/B) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| APC | |
| Human | |
| Primary |
| HIR2 (GA-R2) | |
| Monoclonal | |
| Mouse | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title