Learn More
Epredia™ CD24 (GPI-Linked Surface Mucin) Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures.
Brand: Epredia™ MS-1279-PABX
Additional Details : Weight : 4.08230kg
Description
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. It is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation.Host Species: Mouse
Clone: SN3b
Isotype: IgM/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line
Molecular Weight: 30-70 kDa
Positive Control: Breast CA, Ovarian CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Note:
USA: IVD; Int'l: IVD
Specifications
CD24 (GPI-Linked Surface Mucin) Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
B-Cell | |
Human | |
IgM κ |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
SN3b | |
Unconjugated | |
Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line | |
RUO | |
Primary | |
Concentrated |