Learn More
CD24 Rat, anti-Mouse, PE, Clone: M1/69, STEMCELL Technologies
Rat Monoclonal Antibody
Brand: Stemcell Technologies 60099PE
Description
- The M1/69 antibody reacts with murine CD24, an ∼35 - 45 kDa glycoprotein expressed on lymphocytes, granulocytes, thymocytes, erythrocytes, epithelial cells, neurons, and dendritic cells
- CD24 is differentially expressed during T and B cell development, and is not found on the majority of mature peripheral T cells or plasma cells
- It is expressed on pro-B, pre-B and mature B cells, but expression is strongly down-regulated after B cell activation
- CD24 is involved in regulating B cell apoptosis and in preventing terminal differentiation of B cells into antibody-secreting plasmablasts and plasma cells
- It acts as an adhesion or co-stimulatory molecule and is involved in lymphocyte activation, proliferation and differentiation through homophilic interactions or by binding to ligands such as P-selectin (CD62P)
Specifications
| CD24 | |
| Monoclonal | |
| PE | |
| 200 μg | |
| Mouse | |
| B Cells, Mammary Cells |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| M1/69 | |
| Rat | |
| Primary | |
| IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.