Learn More
CD24 Rat, anti-Mouse, PE, Clone: M1/69, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60099PE
Encompass_Preferred
 View more versions of this product
Product Code. 17149001
  • 444.15€ / 200µg
Description
  • The M1/69 antibody reacts with murine CD24, an ∼35 - 45 kDa glycoprotein expressed on lymphocytes, granulocytes, thymocytes, erythrocytes, epithelial cells, neurons, and dendritic cells
  • CD24 is differentially expressed during T and B cell development, and is not found on the majority of mature peripheral T cells or plasma cells
  • It is expressed on pro-B, pre-B and mature B cells, but expression is strongly down-regulated after B cell activation
  • CD24 is involved in regulating B cell apoptosis and in preventing terminal differentiation of B cells into antibody-secreting plasmablasts and plasma cells
  • It acts as an adhesion or co-stimulatory molecule and is involved in lymphocyte activation, proliferation and differentiation through homophilic interactions or by binding to ligands such as P-selectin (CD62P)
Specifications
Specifications
CD24
Monoclonal
PE
200 μg
Mouse
B Cells, Mammary Cells
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
M1/69
Rat
Primary
IgG2b κ