Learn More
CD25/IL-2Rα Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunofluorescence and flow cytometry experiments with Epredia™ CD25/IL-2Rα Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-203-P0
Description
IL-2R (T-cell growth factor receptor) consists of a chain of 55kDa (also known as Tac antigen or p55 or α chain) and a p75 or Β chain. IL-2Rα is over-expressed on the surface of activated T-cells, HTLV-1 or -2 positive T-cell lines, and moderately expressed on activated B-cells, NK-cells, and macrophages.Host Species: Mouse
Clone: IL2R.1
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant human interleukin-2 receptor
Molecular Weight: 55 kDa
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
| CD25/IL-2Ralpha Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| B-Cell | |
| Human | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence | |
| IL2R.1 | |
| Unconjugated | |
| Recombinant human interleukin-2 receptor | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.