missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Mouse, anti-Human, APC, Clone: BC96, STEMCELL Technologies
Mouse Monoclonal Antibody
82.35€ - 209.25€
Specifications
| Antigen | CD25 |
|---|---|
| Clone | BC96 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17189201
|
Stemcell Technologies
60158AZ.1 |
25 Tests |
82.35€
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17179201
|
Stemcell Technologies
60158AZ |
100 Tests |
209.25€
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The BC96 antibody reacts with human CD25, a 55 kDa type I integral transmembrane glycoprotein
- CD25 is involved in the activation and proliferation of lymphocytes and can be used in the identification of T cell subsets in viral infections like HIV and HTLV
- CD25 is present on activated T and B cells, monocytes, macrophages, and regulatory T cells
- Expression of CD25, together with CD4 and FOXP3, is considered a phenotypic signature for regulatory T cells
- CD25 has a low affinity for its IL-2 ligand but associates with IL-2 receptor β (CD122) and common γ (CD132) chains to form a high affinity IL-2 receptor
Specifications
| CD25 | |
| Flow Cytometry | |
| APC | |
| Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| Primary |
| BC96 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title