Learn More
CD25 Mouse, anti-Human, FITC, Clone: BC96, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60158FI.1
Description
- The BC96 antibody reacts with human CD25, a 55 kDa type I integral transmembrane glycoprotein
- CD25 is involved in the activation and proliferation of lymphocytes and can be used in the identification of T cell subsets in viral infections like HIV and HTLV
- CD25 is present on activated T and B cells, monocytes, macrophages, and regulatory T cells
- Expression of CD25, together with CD4 and FOXP3, is considered a phenotypic signature for regulatory T cells
- CD25 has a low affinity for its IL-2 ligand but associates with IL-2 receptor β (CD122) and common γ (CD132) chains to form a high affinity IL-2 receptor
Specifications
| CD25 | |
| Monoclonal | |
| FITC | |
| 25 Tests | |
| Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| B Cells, T Cells, T Cells, Regulatory |
| Flow Cytometry | |
| BC96 | |
| Mouse | |
| Primary | |
| IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.