Learn More
CD25 Mouse, anti-Human, FITC, Clone: BC96, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60158FI.1
Encompass_Preferred
 View more versions of this product
Product Code. 17119211
  • 94.50€ / 25 tests
Description
  • The BC96 antibody reacts with human CD25, a 55 kDa type I integral transmembrane glycoprotein
  • CD25 is involved in the activation and proliferation of lymphocytes and can be used in the identification of T cell subsets in viral infections like HIV and HTLV
  • CD25 is present on activated T and B cells, monocytes, macrophages, and regulatory T cells
  • Expression of CD25, together with CD4 and FOXP3, is considered a phenotypic signature for regulatory T cells
  • CD25 has a low affinity for its IL-2 ligand but associates with IL-2 receptor β (CD122) and common γ (CD132) chains to form a high affinity IL-2 receptor
Specifications
Specifications
CD25
Monoclonal
FITC
25 Tests
Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human
B Cells, T Cells, T Cells, Regulatory
Flow Cytometry
BC96