Learn More
CD25 Rat, anti-Mouse, PE, Clone: 3C7, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60010PE
Encompass_Preferred
 View more versions of this product
Product Code. 17138411
  • 405.00€ / 100µg
Description
  • The 3C7 antibody reacts with murine CD25 (interleukin‐2 receptor α chain; IL‐2Rα), an ∼55 kDa type 1 transmembrane glycoprotein expressed on the surface of T and B cell progenitors and activated mature B and T cells
  • CD25 has been used as a marker to identify CD4+/FoxP3+ regulatory T cells in mice
  • Mutations in CD25 result in severe immunodeficiency, underscoring the importance of the roles the protein plays in the differentiation, activation and proliferation of lymphocytes, and in the maintenance of self-tolerance
  • CD25 per se has low affinity for its IL-2 ligand but forms dimers with CD122 (IL-2Rβ) and CD132 (IL-2Rγ) that together associate to form the IL-2R receptor, which binds IL-2 with high affinity
  • CD25 acts to increase the specificity and affinity of IL-2 binding and is necessary for receptor clustering and signal transduction by the complex
  • Binding of the 3C7 antibody inhibits binding of IL‐2 to free CD25 and the IL-2R receptor
  • The epitope is distinct from those recognized by the anti-CD25 antibody clones 7D4 and PC61
Specifications
Specifications
CD25
Monoclonal
PE
100 μg
Mouse
T Cells, T Cells, Regulatory
CyTOF, Flow Cytometry, Functional Assay, Immunohistochemistry
3C7
Rat
Primary
IgG2b κ