missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ P2Y2 Recombinant Rabbit Monoclonal Antibody (HL2801)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556827
This item is not returnable.
View return policy
Description
P2Y2 Recombinant Monoclonal Antibody for ICC/IF, IHC (P)
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to both adenosine and uridine nucleotides. It may participate in control of the cell cycle of endometrial carcinoma cells. Three transcript variants encoding the same protein have been identified for this gene.
Specifications
| P2Y2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P35383, P41231 | |
| P2RY2 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain mouse of P2Y2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| HL2801 | |
| Unconjugated | |
| P2RY2 | |
| ATP receptor; HP2U; P2RU1; P2ry2; P2U; P2U nucleotide receptor; P2U purinoceptor 1; P2U receptor 1; P2U1; P2UR; P2Y ATP receptor 2; P2Y purinoceptor 2; P2Y2; P2Y2R; P2Y-like receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled 2; purinergic receptor P2Y, G-protein coupled, 2; purinergic receptor P2Y2; purinoceptor P2Y2; R12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18442, 5029 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction