missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD274 / PD-L1 Rat, PE, Clone: 10F.9G2, enQuireBio™
Rat Monoclonal Antibody
131.25€ - 275.10€
Specifications
| Antigen | CD274 / PD-L1 |
|---|---|
| Clone | 10F.9G2 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15922648
|
EnquireBio
QAB95-PE-25ug |
25 μg |
131.25€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15912648
|
EnquireBio
QAB95-PE-100ug |
100 μg |
275.10€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| CD274 / PD-L1 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 60533 | |
| IgG2b κ | |
| Protein A/G Purified |
| 10F.9G2 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| NP_068693 | |
| Cd274 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title