Learn More
CD279 (PD-1) Mouse anti-Human, PE, Clone: J110, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech PE-65119-100TESTS
Product Code. 16821716
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
The CD279 (PD-1) antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD279 (PD-1). This antibody recognizes human antigen. The CD279 (PD-1) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
Specifications
CD279 (PD-1)
Monoclonal
5 μL (1 μg)/Test
Mouse
Primary
Human
Flow Cytometry
J110
PE
100 Tests
5133