missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Antibody (C28/1636), Novus Biologicals™
Mouse Monoclonal Antibody
289.00€ - 542.00€
Specifications
| Antigen | CD28 |
|---|---|
| Clone | C28/1636 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18278720
|
Novus Biologicals
NBP2-53342-20UG |
20 μg |
289.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18189379
|
Novus Biologicals
NBP2-53342-100UG |
100 μg |
542.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD28 Monoclonal specifically detects CD28 in Human samples. It is validated for Flow Cytometry.Specifications
| CD28 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 940 | |
| Recombinant human full-length CD28 protein | |
| Primary | |
| Store at 4C. |
| C28/1636 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
| CD28 | |
| IgG1 | |
| Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title