Learn More
IBA Lifesciences CD3 Fab-TACS™ Agarose Column Starter Kit
Label-free isolation of CD3+ T cells
154.00€ - 265.50€
Product Code | Brand | Target Species | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Target Species | Price | Quantity & Availability | |||||
17217740
|
IBA Lifesciences
6-3201-002 |
Human |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
17277740
|
IBA Lifesciences
6-3304-002 |
Mouse |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Fab-TACS™ - Traceless Affinity Cell Selection - is a positive cell isolation approach based on our Strep-tag™ technology
- CD3 is a co-receptor that associates with the T cell receptor and plays a role in T cell activation
- Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation
- Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix
- Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target cells are efficiently washed away
- In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps
- After elution, the Fab-Streps self-dissociate from the cell surfaces due to their low affinity
- The label-free authentic target cells are now ready for further downstream applications
Specifications
2°C to 8°C |
For Research Use Only.