Learn More
CD3 Molecular Complex Rat anti-Mouse, FITC, Clone: 17A2, BD
Rat Monoclonal Antibody
183.70€ - 455.00€
Specifications
Antigen | CD3 Molecular Complex |
---|---|
Clone | 17A2 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15831349
|
BD Biosciences
561798 |
0.1 mg |
183.70€
Each |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15838038
|
BD Biosciences
555274 |
0.5 mg |
455.00€
Each |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The 17A2 monoclonal antibody specifically binds to the T-cell receptor-associated CD3 complex that is expressed on many thymocytes and mature T lymphocytes. Plate-bound 17A2 antibody has been reported to induce IL-2 production by cultured T cells in the absence of accessory cells. The binding of the 17A2 antibody to T cells can be blocked by the anti-CD3e mAb 145-2C11. This suggests that the 17A2 antibody recognizes an epitope of the CD3 epsilon chain. In vivo treatment with 17A2 antibody has been reported to partially deplete T lymphocytes and temporarily down-modulates CD3 expression on T cells.
Specifications
CD3 Molecular Complex | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified |
17A2 | |
Flow Cytometry | |
FITC | |
RUO | |
Mouse | |
γδ TCR-positive T-T hybridoma D1 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.