Learn More
CD3 Mouse anti-Human, APC, Clone: UCHT1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech APC-65151-100TESTS
Additional Details : Weight : 0.00010kg
Description
The CD3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD3. This antibody recognizes human antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
CD3 | |
Monoclonal | |
5 μL (0.25 μg)/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
UCHT1 | |
APC | |
100 Tests | |
916 | |
IgG1 |