Learn More
Invitrogen™ CD305 (LAIR1) Monoclonal Antibody (NKTA255), PE, eBioscience™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ 12-3059-42
Product Code. 15506966
  • 300.00€ / 100 tests
Description
Description: This NKTA255 monoclonal antibody reacts with human CD305, which is also known as leukocyte-associated Ig-like receptor (LAIR) 1. CD305 is a type I transmembrane glycoprotein that possesses an extracellular Ig-like domain and two intracellular ITIMs. CD305 is expressed on nearly all immune cells, including T, NK, and B cells, as well as monocytes, dendritic cells, eosinophils, basophils, and mast cells. In humans, this molecule is also expressed on CD34+ hematopoietic progenitor cells and thymocytes. Several isoforms of CD305 exist due to alternative splicing, and their expression may differ between cell types. As an inhibitory receptor, CD305 blocks cytokine-mediated signaling, cell proliferation, and apoptosis. Studies have also identified collagens as the ligands for CD305. The NKTA255 antibody has been reported to inhibit NK cytolytic activity. Applications Reported: This NKTA255 antibody has been reported for use in flow cytometric analysis. Applications Tested: This NKTA255 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufactu...
CD305, also known as LAIR1, is a type I transmembrane glycoprotein with an extracellular Ig-like domain and two intracellular ITIMs. It is expressed on a wide range of immune cells, including T, NK, and B cells, monocytes, dendritic cells, eosinophils, basophils, and mast cells. In humans, CD305 is also present on CD34+ hematopoietic progenitor cells and thymocytes. Several isoforms exist due to alternative splicing, with expression varying between cell types. CD305 functions as an inhibitory receptor, playing a key role in regulating immune responses. It blocks cytokine-mediated signaling, cell proliferation, and apoptosis. In NK, B, and T cells, CD305 modulates cytolytic functions and cytokine production, down-regulating IL2 and IFNG in CD4+ T cells while inducing transforming growth factor beta. In B cells, it reduces IgG and IgE production and decreases IL8, IL10, and TNF secretion. Activation of CD305 through tyrosine phosphorylation recruits phosphatases PTPN6 and PTPN11, although it may also function independently of SH2-containing phosphatases. It inhibits differentiation of peripheral blood precursors into dendritic cells and has been shown to induce apoptosis in myeloid leukemia cell lines. Collagens are identified as ligands for CD305, underscoring its role in immune regulation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD305 (LAIR1)
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q6GTX8
LAIR1
Affinity chromatography
RUO
3903
4°C, store in dark, DO NOT FREEZE!
Liquid
Responsibly packaged
Flow Cytometry
NKTA255
PE
LAIR1
5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; Lair-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1
Mouse
100 Tests