Learn More
CD32 Mouse anti-Rat, Unlabeled, Clone: D34-485, BD
Mouse Monoclonal Antibody
246.00€ - 460.00€
Specifications
Antigen | CD32 |
---|---|
Clone | D34-485 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15806208
|
BD Biosciences
550270 |
0.1 mg |
246.00€
Each |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15816208
|
BD Biosciences
550271 |
0.5 mg |
460.00€
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The D34-485 antibody reacts specifically with CD32, the FcγII receptor. Rat CD32 is expressed on B lymphocytes, myeloid cells, and some lymphocytes in the thymic medulla. D34-485 mAb blocks binding of aggregated immunoglobulins to the FcγIi receptors in vitro. This antibody is routinely tested by flow cytometric analysis and in vitro blocking. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: D34-485
Isotype: IgG1 κ
Species Reactivity: Rat
Immunogen: Recombinant Rat CD32 Protein
Blocking Assay, Flow Cytometry
Specifications
CD32 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
Fcgr2b; FcRII; Fc-gamma RII; FcγRII; FcγII receptor |
D34-485 | |
Flow Cytometry | |
Unconjugated | |
RUO | |
Rat | |
Recombinant Rat CD32 Protein | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.