Learn More
CD328 (Siglec7) Monoclonal Antibody (eBioQA79 (QA79)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-5759-95
Description
Description: The eBioQA79 monoclonal antibody reacts with adhesion inhibitory receptor molecule 1 (AIRM1), also known as sialic acid binding immunoglobulin-like lectin 7 (Siglec-7), p75 and CDw328. AIRM1 is a cell surface receptor that belongs to the sialoadhesin family and displays homology with the myeloid cell antigen CD33. p75/AIRM1 expression is confined to NK cells in lymphoid cells. eBioQA79 crosslinking inhibits NK function. p75/AIRM1 is expressed at various stages of myeloid differentiation. Presence of this protein is a useful way to monitor cellular proliferation and determine proliferative states of cells, especially in diseases such as chronic myeloid leukemia. Applications Reported: This eBioQA79 (QA79) antibody has been reported for use in flow cytometric analysis, and immunoprecipitation. Applications Tested: This eBioQA79 (QA79) antibody has been tested by flow cytometric analysis. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
CD328 (Siglec7) Monoclonal antibody specifically detects CD328 (Siglec7) in Human samples. It is validated for Flow Cytometry, Immunoprecipitation
Specifications
| CD328 (Siglec7) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q9Y286 | |
| Mouse | |
| 10 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation | |
| eBioQA79 (QA79) | |
| Unconjugated | |
| SIGLEC7 | |
| Adhesion inhibitory receptor molecule 1; adhesion inhibitory receptor molecule 1, siglec-7; AIRM1; AIRM-1; CD328; CDw328; D-siglec; p75; p75/ AIRM-1; p75/AIRM1; QA79; QA79 membrane protein; sialic acid binding Ig like lectin 7; sialic acid binding Ig-like lectin 7; sialic acid binding immunoglobulin-like lectin 7; sialic acid-binding Ig-like lectin 7; Siglec; SIGLEC19P; SIGLEC7; SIGLEC-7; SIGLECP2 | |
| Affinity chromatography | |
| RUO | |
| 27036 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.