missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (rCR1/8597), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24139AF350
579.70 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD35 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Recombinant fragment (around aa600-900) of human CD35 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Immunology | |
| 1378 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rCR1/8597 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction