missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (To5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
201.00€ - 351.00€
Specifications
| Antigen | CD35 |
|---|---|
| Clone | To5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699646
|
Novus Biologicals
NBP2-50177-0.025ml |
0.025 mL |
201.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657246
|
Novus Biologicals
NBP2-50177 |
0.1 mL |
351.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOFSpecifications
| CD35 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS | |
| 1378 | |
| IgG1 | |
| Protein A purified |
| To5 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| C3b receptor-enriched precipitates from tonsil cell membranes | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title